Learn More
Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts
Product Code. 12413647
Click to view available options
Type:
End Gates
Gasket
Gel Tray
Unit Size:
2 sets
Each
Product Code. 12413647
Brand: Thermo Scientific™ A1EG
Only null left
Add to basket
Add to basket
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
Choose from various Thermo Scientific™ Owl™ A1 Large Gel System Replacement Parts, including gasketed UVT gel trays, replacement gaskets and external casters.
  • UVT gel tray incorporates gasketed end gates which provide a leakproof seal without tape
  • Comb slots are positioned so you can arrange up to 10 sample sets on the same gel
  • Unique electrode placement allows this gel to be run quickly without compromising the resolution
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Owl A1 Large Gel Electrophoresis System
Product Line Owl
Type End Gates
Includes 1 Pair End Gates