missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OVOL2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5115700
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total OVOL2.
DNA-binding protein that binds to the 5'-G[GCT]GGGGG-3' core sequence. Probably acts as a transcription regulator.
Specifications
| OVOL2 | |
| Polyclonal | |
| Unconjugated | |
| OVOL2 | |
| 1700108N11Rik; 1810007D21Rik; EUROIMAGE566589; hOvo2; MOVO; M-OVO; movo2; M-OVO-A; M-OVO-B; MOVO-B zinc finger protein; MOVO-C zinc finger protein; OVO homolog-like 2; ovo like zinc finger 2; Ovo2; OVOL2; ovo-like 2; ovo-like 2 (Drosophila); ovo-like zinc finger 2; PPCD1; transcription factor Ovo-like 2; Zfp339; zinc finger OVO2; Zinc finger protein 339; zinc finger protein mOVO; Znf339 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 107586, 296201, 58495 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q8CIV7, Q9BRP0 | |
| OVOL2 | |
| A synthesized peptide derived from human OVOL2(Accession Q9BRP0), corresponding to amino acid residues V200-L250. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction