missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OVOL2 Mouse anti-Human, Clone: PCRP-OVOL2-2A1, Novus Biologicals™
Mouse Monoclonal Antibody
£196.00
Specifications
| Antigen | OVOL2 |
|---|---|
| Clone | PCRP-OVOL2-2A1 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array |
| Applications | Flow Cytometry, Immunofluorescence, Peptide Array |
Description
OVOL2 Monoclonal specifically detects OVOL2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Protein Array.Specifications
| OVOL2 | |
| 0.2 mg/ml | |
| Flow Cytometry, Immunofluorescence, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| bA504H3.3, hOvo2, ovo-like 2 (Drosophila), transcription factor Ovo-like 2, Zinc finger protein 339EUROIMAGE566589, ZNF339 | |
| Recombinant full-length human OVOL2 protein | |
| Primary | |
| Store at 4°C. |
| PCRP-OVOL2-2A1 | |
| Flow Cytometry 1-2 ug/million cells, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 58495 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title