missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OVOL2 Antibody (PCRP-OVOL2-2A1), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-08926AF594
This item is not returnable.
View return policy
Description
OVOL2 Monoclonal antibody specifically detects OVOL2 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array
Specifications
| OVOL2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| bA504H3.3, hOvo2, ovo-like 2 (Drosophila), transcription factor Ovo-like 2, Zinc finger protein 339EUROIMAGE566589, ZNF339 | |
| Recombinant full-length human OVOL2 protein (Uniprot: Q9BRP0) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Protein Array | |
| PCRP-OVOL2-2A1 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 58495 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction