missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ OVOL2 Antibody (OVOL2/6776), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-26720-20ug
This item is not returnable.
View return policy
Description
OVOL2 Monoclonal antibody specifically detects OVOL2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| OVOL2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT | |
| bA504H3.3, hOvo2, ovo-like 2 (Drosophila), transcription factor Ovo-like 2, Zinc finger protein 339EUROIMAGE566589, ZNF339 | |
| Recombinant fragment (around aa175-275) of human OVOL2 protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| OVOL2/6776 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 58495 | |
| Store at 4°C. | |
| IgG2 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction