missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OVOL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14082-1-AP-150UL
This item is not returnable.
View return policy
Description
OVOL1 is a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.This gene encodes a protein highly similar to Drosophila and mouse proteins. In Drosophila the ovo protein plays a critical role in Drosophila oogenesis and cuticle formation. In mice the ovo like protein is involved in hair formation and spermatogenesis. The function of the human gene product has not been determined.Specifications
| OVOL1 | |
| Polyclonal | |
| Unconjugated | |
| OVOL1 | |
| HOVO1, ovo like 1(Drosophila), OVOL1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18426, 309164, 5017 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O14753, Q9WTJ2 | |
| Ovol1 | |
| OVOL1 Fusion Protein Ag5210 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction