missing translation for 'onlineSavingsMsg'
Learn More

OVGP1 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18400473 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18400473 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18400473 Supplier Novus Biologicals Supplier No. NBP176939AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OVGP1 Polyclonal antibody specifically detects OVGP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen OVGP1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias CHIT5, EGP, Estrogen-dependent oviduct protein, MUC9, mucin 9, mucin-9, OGP, oviduct glycoprotein, Oviductal glycoprotein, oviductal glycoprotein 1, 120kDa, oviductin, oviduct-specific glycoprotein
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human OVGP1. The immunogen is located within amino acids 60 - 110 of OVGP1. Amino Acid Squence: ILYPEFNKLKERNRELKT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5016
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.