Learn More
Chemsplash Overshoes (Pairs) PB 6B Elasticated Ankle
Product Code. 30058378
Change view
Click to view available options
Size:
Large
X-Large
Unit Size:
200 pairs
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
30058378 X-Large 200 pairs
30058391 Large 200 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30058378 Supplier Chemsplash Supplier No. 2560.400.007
Estimated Shipment: 02-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust.
Chemsplash Elasticated Overshoe
  • Chemsplash 55GSM Type PB 6B Material
  • 16 in. in Length
  • Silicone and Latex Free
  • Non-Linting
  • Anti-Static Material
  • Conforms to ISO 14644-1:2015 Cleanroom 5 - 9
  • PFAS Free
Description: The Chemsplash Elasticated Overshoe is made from Chemsplash soft and microporous fabric. It is strong and resistant to chemical splash and dust. It is anti-static to meet EN1149-5:2008, lint-free, and suitable for cleanroom applications. Conforms to ISO 14644-1:2015 Cleanroom 5 - 9. Chemsplash accessories can be used to protect the wearer from their environment or the environment from the wearer. The range has been designed to allow for maximum comfort and wearability.
Specifications
Size X-Large
Material Microporous Fabric
Latex Free Yes
Color White
Disposable Single-use
Sterility Non-sterile
Closure Type Elastic Ankle
Sole Type Seamless
Resistance Liquid
Product Type Controlled Environments Shoe Covers
Cleanroom Class Class 100-1000 (ISO 5-6)
Show More Show Less
Name des Produkts
Wählen Sie ein Thema
Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.