missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Overlay Sheet

Product Code. 11739665
Click to view available options
For Use With (Equipment):
Fisherbrand GD-4534 Gel Dryer System
Fisherbrand GD-5040 Gel Dryer System
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11739665

Brand: Fisherbrand™ VGDSRO

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Accessory for use with vacuum gel dryers.

  • Translucent
  • Silicone rubber construction
  • Allows gels to be visualised during drying process

Specifications

Color Translucent
For Use With (Equipment) Fisherbrand GD-4534 Gel Dryer System
Type Overlay Sheet
Material Silicone Rubber
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.