Learn More
BINDER™ Overheating Alarm
Shop All BINDER Products
Click to view available options
Material:
Plastic
Unit Size:
Each
Description
- Audible
- Can be switched off
- Adjustable limit on the independent temperature safety device
- Switchable acoustic alarm
Specifications
Specifications
| Description | Alarm function for overheating - Switchable acoustic alarm, with adjustable limit value on the independent temperature safety device |
| For Use With (Equipment) | BINDER Chambers BF 56, BF 115, BF 260, BF 720 |
| Material | Plastic |
| Type | Alarm |