missing translation for 'onlineSavingsMsg'
Få mere at vide

R&D Systems™ Ovary Tissue Slides (Papillary Serous Cystadenocarcinoma)- Paraffin

Artikelnummer. 18726163 Shop alle Bio Techne produkter
Skift visning
Klik for at se tilgængelige muligheder
Species:
Human
Pakningsstørrelse:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Species Pakningsstørrelse
18726163 Human Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 18726163 Leverandør R&D Systems™ Leverandørnr. NBP230296
  • ONLINE EXCLUSIVE PRICE
    £254.99
    £215.25 / Pack of 5
    Save £39.74
    15% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Læg i indkøbskurv
Læg i indkøbskurv

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

A Novus product, part of R&D Systems.

These slides are paraffin coated to prevent sample oxidization, it is recommended that slides are first de-paraffinized by baking at 60 degrees C for 1 hour in a vertical orientation prior to performing antigen retrieval procedures.

TRUSTED_SUSTAINABILITY

Tekniske data

Cross Reactivity Human
Type Ovary Tissue Slides (Papillary Serous Cystadenocarcinoma)
Regulatory Status This product is for research use only and is not approved for use in humans or in clinical diagnosis.
Species Human
Tissue Condition Papillary Serous Cystadenocarcinoma
Product Line Novus
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.