missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ovalbumin Antibody (2C6), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62809AF594
This item is not returnable.
View return policy
Description
Ovalbumin Monoclonal specifically detects Ovalbumin in Chicken samples. It is validated for ELISA.
Specifications
| Ovalbumin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| OVAL | |
| Spleen cells from Ovalbumin immunized Balb/c mice were fused with cells of the mouse myeloma, P3U1 | |
| 0.1 mL | |
| 396058.0 | |
| Chicken | |
| IgE |
| ELISA | |
| 2C6 | |
| ELISA | |
| Allergen Gal d 2, CAP 2, CAP2, Cytoplasmic antiproteinase 2, Egg albumin, Gal d II, Hen egg white, PI8, Plakalbumin, Protease inhibitor 8, Serpin B8, serpin peptidase inhibitor clade B member 8, SERPINB14, SERPINB8 | |
| Mouse | |
| Protein G purified | |
| Primary | |
| NB100-62809 is a mouse monoclonal antibody that specifically recognises ovalbumin (OVA). | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction