missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Kartell™ Oval Stirring Bars
Description
- Ideal for containers or bottles with round bottom
- Suitable for foodstuff
- magnet with PTFE coating
- Oval shape is ideal for ensuring homogeneous mixing in round-bottomed beakers
Specifications
Specifications
| For Use With (Equipment) | Ideal for containers/ bottles with round bottoms |
| Diameter (Metric) | 16 mm |
| Length (Metric) | 35 mm |
| Material | Magnetic Alloy, PTFE |
| Quantity | 10/Pk. |
| Shape | Oval |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?