missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ Oval Stirring Bars

Product Code. 15865537
Change view
Click to view available options
Length (Metric):
20 mm
25 mm
35 mm
Unit Size:
Pack of 10
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
15865537 20 mm Pack of 10
15875537 25 mm Pack of 10
15885537 35 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15865537 Supplier Kartell™ Supplier No. 0068300
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Provide effective stirring of samples via magnetic manipulation

  • Ideal for containers or bottles with round bottom
  • Suitable for foodstuff
  • magnet with PTFE coating
  • Oval shape is ideal for ensuring homogeneous mixing in round-bottomed beakers

Specifications

For Use With (Equipment) Ideal for containers/ bottles with round bottoms
Coating PTFE
Diameter (Metric) 10 mm
Length (Metric) 20 mm
Material Magnetic Alloy, PTFE
Quantity 10/Pk.
Shape Oval
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.