missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ Oval PTFE Stir Bar

Product Code. 10160643 Shop All Fisherbrand Products
Change view
Click to view available options
Length (Metric):
70 mm
100 mm
150 mm
Diameter (Metric):
25 mm
30 mm
40 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) unitSize
10160643 100 mm 30 mm Each
10251952 70 mm 25 mm Each
10464783 150 mm 40 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10160643
Only null left
Add to basket
Add to basket

Provides stable stirring action. These PTFE magnetic stir bars are autoclavable. Provides optimal turbulence for round-bottom flasks and containers.

  • Stable, general-purpose magnetic stir bar
  • Provides excellent turbulence for round-bottom flasks and containers
  • Autoclavable

Specifications

Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Round bottom flasks
Diameter (Metric) 30 mm
Length (Metric) 100 mm
Quantity 1/Ea.
Shape Oval
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.