missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Monoclonal Antibody (OC2-1C6), Invitrogen™
Rat Monoclonal Antibody
Brand: Invitrogen MA516137
Code : LD
Additional Details : Weight : 0.10000kg
Description
Facultative liver stem cell activation was originally described in the rat, where chemical damage and partial hepatectomy caused the emergence of a new cell type, the oval cell. These cells are thought to be bipotential progenitors capable of yielding both hepatocytes and bile ducts. To properly determine the molecular regulation of the progenitor process, appropriate markers are needed to identify and isolate cells that emerge during the oval cell response.Specifications
Oval Cell Marker | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Nonparenchymal cells from DDC-treated mice to induce the oval cell response. | |
RUO | |
Mouse | |
Antibody | |
IgG1 |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Frozen), Western Blot | |
OC2-1C6 | |
Unconjugated | |
Rat | |
100 μL | |
Primary | |
4° C, do not freeze | |
Liquid |