missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Antibody (OC2-2F3) [Janelia Fluor™ 585], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-18964JF585
This item is not returnable.
View return policy
Description
Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Mouse samples. It is validated for Flow Cytometry,Immunohistochemistry,Immunohistochemistry (Frozen)
Specifications
| Oval Cell Marker | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Rat | |
| Tissue culture supernatant | |
| RUO | |
| Mouse | |
| Supernatant |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| OC2-2F3 | |
| 50mM Sodium Borate | |
| Nonparenchymal cells from DDC-treated mice | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction