missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Antibody (MIC1-1C3), PE/Atto594, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP1-18961PEATT594
This item is not returnable.
View return policy
Description
Oval Cell Marker Monoclonal specifically detects Oval Cell Marker in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| Oval Cell Marker | |
| Monoclonal | |
| PE/ATTO 594 | |
| PBS with 0.05% Sodium Azide | |
| Nonparenchymal cells from DDC-treated mice | |
| 0.1 mL | |
| Specific for hepatic proliferating duct cells. | |
| Store at 4°C in the dark. Do not freeze. |
| Flow Cytometry | |
| MIC1-1C3 | |
| Flow Cytometry | |
| Rat | |
| Protein G purified | |
| Primary | |
| Human, Mouse | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction