missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Oval Cell Marker Antibody (MIC1-1C3), Janelia Fluor™ 646, Novus Biologicals™
Shop All Bio Techne ProductsDescription
Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
Specifications
Specifications
| Antigen | Oval Cell Marker |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Classification | Monoclonal |
| Clone | MIC1-1C3 |
| Conjugate | Janelia Fluor 646 |
| Formulation | 50mM Sodium Borate |
| Host Species | Rat |
| Immunogen | Nonparenchymal cells from DDC-treated mice |
| Purification Method | Protein G purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?