missing translation for 'onlineSavingsMsg'
Learn More

Oval Cell Marker Antibody (MIC1-1C3), DyLight 755, Novus Biologicals™

Product Code. 18407252 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407252 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407252 Supplier Novus Biologicals Supplier No. NBP118961IR

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rat Monoclonal Antibody

Oval Cell Marker Monoclonal antibody specifically detects Oval Cell Marker in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oval Cell Marker
Applications Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone MIC1-1C3
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Host Species Rat
Immunogen Nonparenchymal cells from DDC-treated mice
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Hepatic Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.