Learn More
Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
£1376.00 - £3350.00
Specifications
| Antigen | OVA257-264 (SIINFEKL) peptide bound to H-2Kb |
|---|---|
| Clone | eBio25-D1.16 (25-D1.16) |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17180672
|
Invitrogen™
14-5743-37 |
2 mg |
£1376.00
2mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17190672
|
Affymetrix eBioscience
14-5743-95 |
10 mg |
£3350.00
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The 25-D1.16 MAb reacts with the ovalbumin-derived peptide SIINFEKL bound to H-2Kb of MHC class I, but not with unbound H-2Kb, or H-2Kb bound with an irrelevant peptide. This antibody has proven to be very useful tracking the quantity and localization of these specific antigen-presenting cells (APC) in vivo.
OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry.Specifications
| OVA257-264 (SIINFEKL) peptide bound to H-2Kb | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
| eBio25-D1.16 (25-D1.16) | |
| Flow Cytometry, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Mouse | |
| H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb | |
| Primary | |
| 4°C |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.