missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal Antibody (eBio25-D1.16 (25-D1.16)), Brilliant Ultra Violet™ 737, eBioscience™, Invitrogen™

Product Code. 17942131
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17942131 25 μg 25µg
17982141 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17942131 Supplier Invitrogen™ Supplier No. 367574380

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

OVA257-264 (SIINFEKL) peptide bound to H-2Kb Monoclonal antibody specifically detects OVA257-264 (SIINFEKL) peptide bound to H-2Kb in Mouse samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen OVA257-264 (SIINFEKL) peptide bound to H-2Kb
Applications Flow Cytometry
Classification Monoclonal
Clone eBio25-D1.16 (25-D1.16)
Concentration 0.2 mg/mL
Conjugate Brilliant Ultraviolet 737
Formulation PBS with BSA and 0.09% sodium azide; pH 7.2
Gene Alias H-2Kb-SIINFEKL; OVA257-264; OVA257-264 H-2Kb; OVA-Kb
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.