missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OV-6 Antibody (OV-6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB2020CL1
This item is not returnable.
View return policy
Description
OV-6 Monoclonal antibody specifically detects OV-6 in Human,Rat samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF
Specifications
| OV-6 | |
| Monoclonal | |
| CoraFluor 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human, Rat | |
| Purified |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| OV-6 | |
| PBS | |
| Carcinogen-treated rat nodular hepatocytes | |
| 0.1 mL | |
| Primary | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction