missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ OUL 35

Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
Description
OUL 35 is a selective PARP-10 (also known as ADP-ribosyltransferase Diptheria Toxin-like 10, ARTD10) inhibitor (IC50 = 0.329 μM). Displays selectivity for PARP-10 over a panel of PARP family members. Rescues HeLa cells from PARP-10-induced cell death. Sensitizes cancer cells to hydroxyurea-induced genotoxic stress.
Specifications
Specifications
| CAS | 6336-34-1 |
| Synonym | 4,4′-Oxybis[benzamide] |
| Quantity | 5 mg |
| Molecular Formula | C14H12N2O3 |
| Formula Weight | 256.26 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction