missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Ouabain octahydrate, 96%

Product Code. 10154030 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
25g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10154030 1 g Glass bottle 1g
10165353 250 mg Glass bottle 250mg
10771431 5 g Glass bottle 5g
10287353 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10154030 Supplier Thermo Scientific Chemicals Supplier No. 161730010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ouabain octahydrate, CAS # 11018-89-6, (synonym: gamma-Strophanthin), is a plant-derived organic compound that inhibits the Na+/K+ ATPase pump present in all animal cell membranes. | CAS: 11018-89-6 | C29H44O12·8H2O | 728.78 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General description

• Ouabain octahydrate is a water-soluble, light-sensitive powder that is stable at ambient temperatures
• It is a glycoside comprised of rhamnose and ouabagenin, and is derived from the ripe seeds of Strophanthus gratus and the bark of Acokanthera ouabaio

Applications

• Ouabain octahydrate works by binding the Na+/K+ ATPase pump, thus disrupting its function, and leading to an increase in intracellular Na+.
• The inhibitory action of ouabain octahydrate on the Na+/K+ pump in cardiac cells makes it a pharmaceutical cardioactive agent
• Ouabain octahydrate has also been found to induce cytotoxic effects through a steroidal signal transduction pathway, in laboratory preparations

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11018-89-6
Molecular Formula C29H44O12·8H2O
Molecular Weight (g/mol) 728.78
MDL Number MFCD00149240
InChI Key TYBARJRCFHUHSN-DMJRSANLSA-N
Synonym ouabain octahydrate, strophantine octahydrate, quabain octahydrate, ouabain usp, penta a phenanthren-17-yl-2h-furan-5-one;octahydrate, gamma-strophanthin, 3-1r,3s,5s,8r,9s,10r,11r,13r,14s,17r-1,5,11,14-tetrahydroxy-10-hydroxymethyl-13-methyl-3-2r,3r,4r,5r,6s-3,4,5-trihydroxy-6-methyloxan-2-yl oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1h-cyclo, prestwick_370, acocantherine;g-strophanthin
PubChem CID 6364534
IUPAC Name 3-[(1R,3S,5S,8R,9S,10R,11R,13R,14S,17R)-1,5,11,14-tetrahydroxy-10-(hydroxymethyl)-13-methyl-3-[(2R,3R,4R,5R,6S)-3,4,5-trihydroxy-6-methyloxan-2-yl]oxy-2,3,4,6,7,8,9,11,12,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]-2H-furan-5-one;octahydrate
SMILES CC1C(C(C(C(O1)OC2CC(C3(C4C(CCC3(C2)O)C5(CCC(C5(CC4O)C)C6=CC(=O)OC6)O)CO)O)O)O)O.O.O.O.O.O.O.O.O

Specifications

Melting Point 260.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 95% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Beilstein 15, V,5, 123
Merck Index 14, 6901
Specific Rotation Condition − 33.00 (20.00°C c=1,H2O,on dry subs.)
Specific Rotation − 33.00
Solubility Information (1% in water) Clear colorless
Formula Weight 728.78
Percent Purity 96%
Physical Form Crystalline Powder
Chemical Name or Material Ouabain octahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Ouabain octahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H331-Toxic if inhaled.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Ouabain octahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.