Learn More
Invitrogen™ OTX2 Recombinant Rabbit Monoclonal Antibody (14H14L5)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ 701948
Description
This antibody is predicted to react with Rat, Mouse, Pig and Sheep. Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire.
OTX2 is a 289 amino acid containing protein encoding a member of the bicoid sub-family of homeodomain-containing transcription factors. It has two known alternative splice variants with distinct isoforms though presence of other isoforms cannot be excluded. The protein is an early marker of endodermal differentiation of embryonic stem cells and may play a role in the development of the brain and the sensory organs. The proteinmay bind to BCD target sequence (BTS): 5-'TCTAATCCC-3'. Defects in OTX2 are the cause of microphthalmia syndromic type 5 (MCOPS5).
Specifications
| OTX2 | |
| Recombinant Monoclonal | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| P32243, P80206, Q64201 | |
| OTX2 | |
| Peptides corresponding to Human OTX2 (aa 20-39, 276-289). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot, Immunocytochemistry | |
| 14H14L5 | |
| Unconjugated | |
| OTX2 | |
| CPHD6; E130306E05Rik; homeobox Otx2; homeobox protein OTX2; MCOPS5; orthodenticle homeobox 2; orthodenticle homolog 2; OTX; OTX2 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18424, 305858, 5015 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.