missing translation for 'onlineSavingsMsg'
Learn More

Otx2 Mouse anti-Human, Clone: PCRP-OTX2-1E10, Novus Biologicals™

Product Code. 18338225 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18338225 20 μg 20µg
18310127 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18338225 Supplier Novus Biologicals Supplier No. NBP31398620UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Otx2 Monoclonal antibody specifically detects Otx2 in Human samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation
TRUSTED_SUSTAINABILITY

Specifications

Antigen Otx2
Applications Flow Cytometry, ELISA, Immunoprecipitation
Classification Monoclonal
Clone PCRP-OTX2-1E10
Concentration 0.2 mg/ml
Conjugate Unconjugated
Dilution Flow Cytometry 1-2 ug/million cells, ELISA, Immunoprecipitation 1-2 ug/100-500 ug protein
Formulation 10 mM PBS with 0.05% BSA
Gene Accession No. P32243
Gene Alias homeobox protein OTX2, MCOPS5, MGC45000, orthodenticle homeobox 2, Orthodenticle homolog 2, orthodenticle homolog 2 (Drosophila), OTX2
Host Species Mouse
Immunogen Recombinant human full-lengthOtx2protein (Uniprot: P32243)
Purification Method Protein A or G purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers, Vision
Primary or Secondary Primary
Gene ID (Entrez) 5015
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.