missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD7B Monoclonal Antibody (OTI2E2)

OTUD7B Monoclonal Antibody (OTI2E2)
Brand: Invitrogen MA527168
This item is not returnable.
View return policy
Description
Specifications
| OTUD7B | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6GQQ9 | |
| OTUD7B | |
| Human recombinant protein fragment corresponding to amino acids 449-729 of OTUD7B produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2E2 | |
| Unconjugated | |
| OTUD7B | |
| 2900060B22Rik; 4930463P07Rik; AI462125; cellular zinc finger anti-NF-kappaB Cezanne; cellular zinc finger anti-NF-kappa-B protein; CEZANNE; OTU deubiquitinase 7B; OTU domain containing 7B; OTU domain-containing protein 7B; OTUD7B; RGD1308111; Trabid2; TRAF binding domain containing 2; Za20d1; Zinc finger A20 domain-containing protein 1; zinc finger protein Cezanne; zinc finger, A20 domain containing 1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 56957 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering