missing translation for 'onlineSavingsMsg'
Learn More

OTUD7B/Cezanne/ZA20D1 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Product Code. 18319851
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µL
25µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319851 100 μg 100µL
18389486 25 μg 25µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18319851 Supplier Bio-Techne Supplier No. NBP317748100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OTUD7B/Cezanne/ZA20D1 Polyclonal antibody specifically detects OTUD7B/Cezanne/ZA20D1 in Human samples. It is validated for Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD7B/Cezanne/ZA20D1
Applications Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml
Formulation PBS, pH 7.2, 40% glycerol
Gene Alias cellular zinc finger NF-kappaB Cezanne, Cellular zinc finger NF-kappa-B protein, CEZANNEA20 domain containing 1, EC 3.4.19.12, OTU domain containing 7B, OTU domain-containing protein 7B, ZA20D1, Zinc finger A20 domain-containing protein 1, Zinc finger protein Cezanne
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: QGEGKFIFVGTLKMGHRHQYQEEMIQRYLSDAEERFLAEQKQKEAERKIMNGGIGGGPPPAKKPEPDA
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Mucosal Immunology
Primary or Secondary Primary
Gene ID (Entrez) 56957
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.