missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTUD7B/Cezanne/ZA20D1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-35742-100ul
This item is not returnable.
View return policy
Description
OTUD7B/Cezanne/ZA20D1 Polyclonal antibody specifically detects OTUD7B/Cezanne/ZA20D1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/Immunofluorescence
Specifications
| OTUD7B/Cezanne/ZA20D1 | |
| Polyclonal | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| cellular zinc finger NF-kappaB Cezanne, Cellular zinc finger NF-kappa-B protein, CEZANNEA20 domain containing 1, EC 3.4.19.12, OTU domain containing 7B, OTU domain-containing protein 7B, ZA20D1, Zinc finger A20 domain-containing protein 1, Zinc finger protein Cezanne | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 530-700 of human OTUD7B/Cezanne/ZA20D1 (NP_064590.2).,, Sequence:, SKPGGVGTGLGGSSGTETLEKKKKNSLKSWKGGKEEAAGDGPVSEKPPAESVGNGGSKYSQEVMQSLSILRTAMQGEGKFIFVGTLKMGHRHQYQEEMIQRYLSDAEERFLAEQKQKEAERKIMNGGIGGGPPPAKKPEPDAREEQPTGPPAESRAMAFSTGYPGDFTIPR | |
| 100 μL | |
| Mucosal Immunology | |
| 56957 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| ELISA, Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction