missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OTUD6A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562772
This item is not returnable.
View return policy
Description
Immunogen sequence: HRQELEKFQD DSSIESVVED LAKMNLENRP PRSSKAHRKR ERMESEERER QESIFQAEMS EHLAGFKREE E Highest antigen sequence identity to the following orthologs: Mouse - 53%, Rat - 51%.
Deubiquitinating enzymes (DUBs; see MIM 603478) are proteases that specifically cleave ubiquitin linkages, negating the action of ubiquitin ligases. DUBA2 belongs to a DUB subfamily characterized by an ovarian tumor (OTU) domain.
Specifications
| OTUD6A | |
| Polyclonal | |
| Unconjugated | |
| OTUD6A | |
| DUBA2; DUBA-2; EG408193; Hin6; Hin-6 protease; HSHIN6; OTU deubiquitinase 6A; OTU domain containing 6A; OTU domain-containing protein 6A; OTUD6A; RGD1560855 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 139562 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7L8S5 | |
| OTUD6A | |
| Recombinant protein corresponding to Human OTUD6A. Recombinant protein control fragment (Product #RP-99994). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction