missing translation for 'onlineSavingsMsg'
Learn More

OTUD5 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Product Code. 18423014 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423014 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423014 Supplier Novus Biologicals Supplier No. NBP176943AF532

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

OTUD5 Polyclonal antibody specifically detects OTUD5 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD5
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 532
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme A, DKFZp761A052, DUBAdeubiquinating enzyme A, EC 3.4.19.12, MGC104871, OTU domain containing 5, OTU domain-containing protein 5
Host Species Rabbit
Immunogen Antibody was raised against a 13 amino acid synthetic peptide near the carboxy terminus of the human OTUD5. The immunogen is located within amino acids 400 - 450 of OTUD5. Amino Acid Squence: VRGPSQPKASAT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 55593
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.