missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OTUD4 Polyclonal Antibody

Product Code. 12828852
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12828852 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12828852 Supplier Invitrogen™ Supplier No. PA520707

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is Daudi cell lysate. PA5-20707 can be used with blocking peptide PEP-0822.

OTUD4 alternatively spliced transcript variants have been found for this gene. The smaller protein isoform encoded by the shorter transcript variant is found only in HIV-1 infected cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OTUD4
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene OTUD4
Gene Accession No. B2RRE7, Q01804
Gene Alias 4930431L18Rik; AI449692; D8Ertd69e; DUBA6; HIN1; HIN-1; HIV-1 induced protein HIN-1; HIV-1-induced protein HIN-1; HSHIN1; KIAA1046; mKIAA1046; OTU deubiquitinase 4; OTU domain containing 4; OTU domain-containing protein 4; Otud4; RGD1305606
Gene Symbols OTUD4
Host Species Rabbit
Immunogen An 18 amino acid peptide near the carboxy terminus of human OTUD4.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 307774, 54726, 73945
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.