missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ OTUD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA524508
This item is not returnable.
View return policy
Description
OTUD3 Polyclonal Antibody for IHC (P), Western Blot
Deubiquitinating enzyme that hydrolyzes 'Lys-6'- and 'Lys-11'-linked polyubiquitin. Also hydrolyzes heterotypic (mixed and branched) and homotypic chains.
Specifications
| OTUD3 | |
| Polyclonal | |
| Unconjugated | |
| OTUD3 | |
| DUBA4; KIAA0459; OTU deubiquitinase 3; OTU domain containing 3; OTU domain-containing protein 3; OTUD3; RP11-460G22.1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 23252 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q5T2D3 | |
| OTUD3 | |
| KLH conjugated synthetic peptide between 192-220 amino acids from the central region of human OTUD3. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction