missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Otubain-2 Polyclonal specifically detects Otubain-2 in Human samples. It is validated for ELISA.
Specifications
Specifications
| Antigen | Otubain-2 |
| Applications | ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:5000-1:10000 |
| Formulation | 0.2 um filtered solution in PBS |
| Gene Alias | C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with purified, recombinant Human Otubain-2 (Uniprot#: Q96DC9-1; Met1-His234) |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?