missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-00088-100ul
This item is not returnable.
View return policy
Description
Otubain-2 Polyclonal antibody specifically detects Otubain-2 in Human samples. It is validated for ELISA
Specifications
| Otubain-2 | |
| Polyclonal | |
| ELISA 1:1000-1:2000 | |
| C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2 | |
| Produced in rabbits immunized with purified, recombinant Human Otubain-2 (Uniprot#: Q96DC9-1; Met1-His234) | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 78990 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction