missing translation for 'onlineSavingsMsg'
Learn More

Otubain-2 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18322465 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18322465 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18322465 Supplier Novus Biologicals Supplier No. NBP300089AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Otubain-2 Polyclonal specifically detects Otubain-2 in Human samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Otubain-2
Applications ELISA
Classification Polyclonal
Conjugate Alexa Fluor 700
Formulation 50 mM Sodium Borate
Gene Alias C14orf137chromosome 14 open reading frame 137, Deubiquitinating enzyme OTUB2, EC 3.4.19.12, FLJ21916, MGC3102, OTB2otubain-2, OTU domain, ubiquitin aldehyde binding 2, OTU domain-containing ubiquitin aldehyde-binding protein 2, OTU2ubiquitin thioesterase OTUB2, Otubain-2, ubiquitin-specific protease otubain 2, Ubiquitin-specific-processing protease OTUB2
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Otubain-2 (Q96DC9-1; Met1-His234).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 78990
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.