missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-1 Mouse anti-Human, Clone: 1.70E+11, Novus Biologicals™
Mouse Monoclonal Antibody
£234.00 - £378.00
Specifications
| Antigen | Otubain-1 |
|---|---|
| Clone | 170000000000 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18278391
|
Novus Biologicals
NBP2-59389-50UL |
50 μL |
£234.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18220603
|
Novus Biologicals
NBP2-59389-100UL |
100 μL |
£378.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Otubain-1 Monoclonal specifically detects Otubain-1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
| Otubain-1 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1 | |
| OTUB1 | |
| IgG1 κ | |
| Protein A purified |
| 170000000000 | |
| Western Blot 1:1000, Flow Cytometry 1:10-1:1000, ELISA, Immunocytochemistry/Immunofluorescence 1:100 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Liquid. In PBS (pH 7.4), 10% Glycerol with 0.02% Sodium Azide | |
| 55611 | |
| Recombinant human OTUB1 (1-271aa) purified from E. coli | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title