missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Otubain-1 Antibody (OTI5B8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73166F
This item is not returnable.
View return policy
Description
Otubain-1 Monoclonal specifically detects Otubain-1 in Human, Rat samples. It is validated for Flow Cytometry, Immunohistochemistry.
Specifications
| Otubain-1 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| OTUB1 | |
| Full length recombinant protein of human Otubain-1 (NP_060140) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Rat | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry | |
| OTI5B8 | |
| Flow Cytometry, Immunohistochemistry | |
| Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1 | |
| Mouse | |
| Affinity Purified | |
| Proteases & Other Enzymes | |
| 55611.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction