missing translation for 'onlineSavingsMsg'
Learn More

Otubain-1 Antibody (17E10), DyLight 650, Novus Biologicals™

Product Code. 18430785 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430785 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430785 Supplier Novus Biologicals Supplier No. NBP259389C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Otubain-1 Monoclonal antibody specifically detects Otubain-1 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen Otubain-1
Applications Western Blot, ELISA, Immunofluorescence
Classification Monoclonal
Clone 170000000000
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias Deubiquitinating enzyme OTUB1, EC 3.1.2, EC 3.4.19.12, FLJ20113, FLJ40710, hOTU1, MGC111158, OTB1MGC4584, OTU domain, ubiquitin aldehyde binding 1, OTU domain-containing ubiquitin aldehyde-binding protein 1, OTU1OTU-domain Ubal-binding 1, otubain-1, ubiquitin thioesterase OTUB1, ubiquitin-specific protease otubain 1, Ubiquitin-specific-processing protease OTUB1
Host Species Mouse
Immunogen Recombinant human Otubain-1 (1-271aa) purified from E. coli
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 55611
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.