missing translation for 'onlineSavingsMsg'
Learn More

OTS964 hydrochloride, MedChemExpress

Product Code. 16487907
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16487907 5 mg 5mg
16485876 100 mg 100mg
16473306 25 mg 25mg
16440166 2 mg 2mg
16435188 10 mg 10mg
16431407 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16487907 Supplier Medchem Express Supplier No. HY12467
 Restricted Item
Estimated Shipment: 16-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress OTS964 hydrochloride is an orally active, high affinity and selective TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor with an IC50 of 28 nM. OTS964 hydrochloride is also a potent inhibitor of the cyclin-dependent kinase CDK11, which binds to CDK11B with a Kd of 40 nM.

OTS964 hydrochloride is an orally active, high affinity and selective TOPK (T-lymphokine-activated killer cell-originated protein kinase) inhibitor with an IC50 of 28 nM. OTS964 hydrochloride is also a potent inhibitor of the cyclin-dependent kinase CDK11, which binds to CDK11B with a Kd of 40 nM.

Specifications

Chemical Name or Material OTS964 hydrochloride
CAS 1338545-07-5
Color White
Purity Grade Notes Research
Molecular Formula C23H25ClN2O2S
Quantity 5 mg
Solubility Information DMSO : ≥ 83.33 mg/mL (194.26 mM) ∣H2O : 1.43 mg/mL (3.33 mM; Need ultrasonic)
SMILES OC1=CC(C)=C2C(C3=C(SC=C3)C(N2)=O)=C1C4=CC=C([C@@H](C)CN(C)C)C=C4.Cl
Molecular Weight (g/mol) 428.97
Formula Weight 428.97
Percent Purity 95.59%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.