missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTP Mouse anti-Human, Rat, Clone: CL11225, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-15136-100UL
This item is not returnable.
View return policy
Description
OTP Monoclonal antibody specifically detects OTP in Human, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| OTP | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.2), 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| CL11225 | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 2 to 10 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| homeobox protein orthopedia, MGC3161, orthopedia homeobox, orthopedia homolog, orthopedia homolog (Drosophila) | |
| This antibody was developed against a recombinant protein. Actual immunogen sequence is proprietary information. | |
| 100 μg | |
| Neuroscience | |
| 23440 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction