Learn More
Invitrogen™ OTP Monoclonal Antibody (CL11225)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA544172
Description
The antibody solution should be gently mixed before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: VKCRLGVGGS DPGGHPGDLA PNSDPVEGAT LLPGEDITTV GSTPASLAVS AKDPDKQPGP QGGPNPSQAG QQQGQQKQKR HRTRFTPAQL NELE Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 100%.
This gene encodes a member of the homeodomain (HD) family. HD family proteins are helix-turn-helix transcription factors that play key roles in the specification of cell fates. This protein may function during brain development.
Specifications
| OTP | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5XKR4 | |
| OTP | |
| Recombinant protein corresponding to Human OTP. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| CL11225 | |
| Unconjugated | |
| OTP | |
| homeobox protein orthopedia; orthopedia homeobox; orthopedia homolog; orthopedia homolog (Drosophila); Otp | |
| Mouse | |
| Protein A | |
| RUO | |
| 23440, 294640 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.