Learn More
OTP Antibody (CL11225) [DyLight 680], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-15136FR
Product Code. 30304746
  • £558.00 / 0.10mL
Description
OTP Monoclonal antibody specifically detects OTP in Human,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
OTP
Monoclonal
DyLight 680
homeobox protein orthopedia, MGC3161, orthopedia homeobox, orthopedia homolog, orthopedia homolog (Drosophila)
This antibody was developed using a recombinant protein derived from Q5XKR4, with the exact immunogen sequence remaining proprietary.
0.1 mL
Neuroscience
23440
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
CL11225
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human, Rat
Purified