missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTOP1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24456-1-AP-20UL
This item is not returnable.
View return policy
Description
Required for normal formation of otoconia in the inner ear. Inhibits P2Y purinoceptors. Modulates calcium homeostasis and influx of calcium in response to extracellular ATP.Specifications
| OTOP1 | |
| Polyclonal | |
| Unconjugated | |
| OTOP1 | |
| OTOP1, otopetrin 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 133060, 21906 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.34 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q7RTM1, Q80VM9 | |
| Otop1 | |
| OTOP1 Fusion Protein Ag20011 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction