missing translation for 'onlineSavingsMsg'
Learn More

Otenaproxesul, MedChemExpress

Product Code. 16446626
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16446626 100 mg 100mg
16442417 50 mg 50mg
16416298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16446626 Supplier Medchem Express Supplier No. HY15028
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Otenaproxesul (ATB-346), an orally active non-steroidal anti-inflammatory drug (NSAID), inhibits cyclooxygenase-1 and 2 (COX-1 and 2). Otenaproxesul possesses antiinflammatory and antinociceptive activities.

Otenaproxesul (ATB-346), an orally active non-steroidal anti-inflammatory drug (NSAID), inhibits cyclooxygenase-1 and 2 (COX-1 and 2). Otenaproxesul possesses antiinflammatory and antinociceptive activities.

Specifications

Chemical Name or Material Otenaproxesul
CAS 1226895-20-0
Color Yellow
Purity Grade Notes Research
Molecular Formula C21H19NO3S
Quantity 100 mg
Synonym ATB-346
Solubility Information DMSO : ≥ 51.6 mg/mL (141.20 mM)
SMILES COC1=CC=C(C=C(C(C)C(OC2=CC=C(C(N)=S)C=C2)=O)C=C3)C3=C1
Molecular Weight (g/mol) 365.45
Formula Weight 365.45
Percent Purity 98.35%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.