missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ OTC Recombinant Rabbit Monoclonal Antibody (6B6N2)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA558166

 View more versions of this product

Product Code. 30282745

  • £363.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: LQAATPKGYE PDASVTKLAE QYAKENGTKL LLTNDPLEAA HGGNVLITDT WISMGQEEEK KKRLQAFQGY QVTMKTAKVA ASDWTFLHCL PRKPEEVDDE VFYSPRSLVF PEAENRKWTI MAVMVSLLTD YSPQLQKPKF.

This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

OTC
Recombinant Monoclonal
1.2 mg/mL
PBS with 0.05% BSA, 50% glycerol and 0.09% sodium azide; pH 7.3
P00480
OTC
Recombinant fusion protein containing a sequence corresponding to amino acids 215-354 of human OTC (NP_000522.3).
100 μL
Primary
Human
Antibody
IgG
ELISA, Immunohistochemistry (Paraffin)
6B6N2
Unconjugated
OTC
AI265390; OCTD; ornithine carbamoyltransferase; ornithine carbamoyltransferase, mitochondrial; Ornithine transcarbamylase; Otc; OTCase; Sf; sparse fur; spf
Rabbit
Protein A
RUO
5009
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.