missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OTC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26470-1-AP-20UL
This item is not returnable.
View return policy
Description
This nuclear gene encodes a mitochondrial matrix enzyme. Missense, nonsense, and frameshift mutations in this enzyme lead to ornithine transcarbamylase deficiency, which causes hyperammonemia. Since the gene for this enzyme maps close to that for Duchenne muscular dystrophy, it may play a role in that disease also.Specifications
| OTC | |
| Polyclonal | |
| Unconjugated | |
| OTC | |
| OCTD, ornithine carbamoyltransferase, Ornithine transcarbamylase, OTC, OTCase | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18416, 25611, 5009 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P00480, P00481, P11725 | |
| Otc | |
| OTC Fusion Protein Ag24186 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction