missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTM1 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99663IR
This item is not returnable.
View return policy
Description
OSTM1 Polyclonal specifically detects OSTM1 in Human samples. It is validated for ELISA.
Specifications
| OSTM1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 28962 | |
| Store at 4°C in the dark. | |
| IgG |
| ELISA | |
| DyLight 755 | |
| GIPN, GLGAIP-interacting protein N terminus, grey-lethal osteopetrosis, HSPC019, OPTB5, osteopetrosis associated transmembrane protein 1, osteopetrosis-associated transmembrane protein 1 | |
| Produced in rabbits immunized with purified, recombinant OSTM1 (NP_054747.2; Met1-Pro284). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction