missing translation for 'onlineSavingsMsg'
Learn More
Learn More
OSTM1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93631-0.1ml
This item is not returnable.
View return policy
Description
OSTM1 Polyclonal antibody specifically detects OSTM1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| OSTM1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| GIPN, GLGAIP-interacting protein N terminus, grey-lethal osteopetrosis, HSPC019, OPTB5, osteopetrosis associated transmembrane protein 1, osteopetrosis-associated transmembrane protein 1 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 32-284 of human OSTM1 (NP_054747.2). ALPFGSSPHRVFHDLLSEQQLLEVEDLSLSLLQGGGLGPLSLPPDLPDLDPECRELLLDFANSSAELTGCLVRSARPVRLCQTCYPLFQQVVSKMDNISRAAGNTSESQSCARSLLMADRMQIVVILSEFFNTTWQEANCANCLTNNSEELSNSTVYFLNLFNHTLTCFEHNLQGNAHSLLQTKNYSEVCKNCREAYKTLSSLYSEMQKMNELENKAEPGTHLCIDVEDAMNITRKLWSRTFNCSVPCSDTVP | |
| 0.1 mL | |
| Cancer, Cell Biology, Cell Cycle and Replication, Cytoskeleton Markers, Signal Transduction | |
| 28962 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction